siRNA Details
Gene Symbol: Dsg1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181680.1 | NP_859008.1 | 10 | 1467 | ||
GenBank mRNA | AY192159.1 | - | 10 | 1467 | ||
AY314983.1 | - | 10 | 1367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
