siRNA Details
Gene Symbol: Plekhh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181073.3 | NP_851418.2 | 11 | 1998 | ||
GenBank mRNA | AK122464.1 | - | 11 | 1967 | ||
AK143387.1 | - | 11 | 1815 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Plekhh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181073.3 | NP_851418.2 | 11 | 1998 | ||
GenBank mRNA | AK122464.1 | - | 11 | 1967 | ||
AK143387.1 | - | 11 | 1815 |