siRNA Details
Gene Symbol: Trim52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198601.3 | NP_941003.1 | 1 | 1049 | ||
GenBank mRNA | AK029521.1 | - | 1 | 1055 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
