siRNA Details
Gene Symbol: Nhlrc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172501.2 | NP_766089.1 | 3 | 412 | ||
GenBank mRNA | AK029465.1 | - | 3 | 407 | ||
AK033167.1 | - | 3 | 412 | |||
AK133986.1 | - | 362 | ||||
BC087903.1 | - | 3 | 399 | 30250042 | ||
BC107356.1 | - | 3 | 400 | 40055875 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
