siRNA Details
Gene Symbol: Ubxn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285928.1 | NP_001272857.1 | 4 | 809 | ||
NM_001285929.1 | NP_001272858.1 | 3 | 698 | |||
NM_001285930.1 | NP_001272859.1 | 3 | 562 | |||
NM_178671.5 | NP_848786.1 | 2 | 431 | |||
GenBank mRNA | AK045964.1 | - | 2 | 389 | ||
AK046949.1 | - | 4 | 810 | |||
AK077656.1 | - | 2 | 433 | |||
AK080662.1 | - | 2 | 397 | |||
AK133185.1 | - | 2 | 417 | |||
AK161245.1 | - | 2 | 418 | |||
AK161296.1 | - | 2 | 394 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
