siRNA Details
Gene Symbol: Cfap44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033247.1 | NP_001028419.1 | 4 | 346 | ||
GenBank mRNA | BC050980.1 | - | 4 | 345 | ||
BC100518.1 | - | 4 | 346 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cfap44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033247.1 | NP_001028419.1 | 4 | 346 | ||
GenBank mRNA | BC050980.1 | - | 4 | 345 | ||
BC100518.1 | - | 4 | 346 |