siRNA Details
Gene Symbol: Rho | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145383.1 | NP_663358.1 | 3 | 685 | ||
GenBank mRNA | AK044333.1 | - | 3 | 705 | ||
AK044412.1 | - | 3 | 703 | |||
BC013125.1 | - | 3 | 685 | 4500760 | ||
BC031766.1 | - | 3 | 686 | 4527040 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
