siRNA Details
Gene Symbol: Zfp273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198322.3 | NP_938081.2 | 4 | 774 | ||
GenBank mRNA | AK078495.1 | - | 4 | 734 | ||
BC094501.1 | - | 4 | 758 | 4952142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
