siRNA Details
Gene Symbol: N4bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145974.3 | NP_666086.2 | 3 | 935 | ||
GenBank mRNA | AK049249.1 | - | 3 | 705 | ||
AK077848.1 | - | 3 | 699 | |||
AK141228.1 | - | 3 | 729 | |||
AK142500.1 | - | 3 | 698 | |||
AK170270.1 | - | 3 | 733 | |||
AK170521.1 | - | 3 | 730 | |||
BC006740.1 | - | 3 | 648 | 3158173 | ||
BC055831.1 | - | 3 | 935 | 4187601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
