siRNA Details
Gene Symbol: Chpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146690.1 | NP_001140162.1 | 4, 5 | 760 | ||
NM_144807.3 | NP_659056.3 | 4, 5 | 760 | |||
GenBank mRNA | AK007964.1 | - | 4, 5 | 680 | ||
AK032497.1 | - | 4, 5 | 655 | |||
AY445814.1 | - | 4, 5 | 655 | |||
EL958096.1 | - | 3, 4 | 360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
