siRNA Details
Gene Symbol: Ddx46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282055.1 | NP_001268984.1 | 17 | 2446 | ||
GenBank mRNA | AK088950.1 | - | 16 | 4036 | ||
AK129220.1 | - | 16 | 4037 | |||
AK134073.1 | - | 5 | 1692 | |||
BC026492.1 | - | 10 | 1391 | 4505095 | ||
BC092240.1 | - | 16 | 2357 | 30662573 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
