siRNA Details
Gene Symbol: Kctd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172509.3 | NP_766097.1 | 4 | 1009 | ||
GenBank mRNA | AK029942.1 | - | 4 | 985 | ||
AK034583.1 | - | 4 | 985 | |||
AK083583.1 | - | 4 | 1010 | |||
AK140606.1 | - | 4 | 984 | |||
AK162568.1 | - | 4 | 1038 | |||
AK178565.1 | - | 1 | 128 | |||
AK195855.1 | - | 1 | 128 | |||
BC051544.1 | - | 3 | 552 | |||
BC139070.1 | - | 4 | 911 | 8862092 | ||
BC141201.1 | - | 4 | 911 | 9055813 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
