siRNA Details
Gene Symbol: Mfsd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114662.1 | NP_001108134.1 | 9, 10 | 1654 | ||
GenBank mRNA | AK085484.1 | - | 6, 7 | 1070 | ||
FN434127.1 | - | 2, 3 | 89 |

More Information
Other Species Targeted

Gene Ontology Categories:
