siRNA Details
Gene Symbol: Zfp583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033249.3 | NP_001028421.1 | 5 | 1178 | ||
GenBank mRNA | AK038614.1 | - | 5 | 959 | ||
AK133380.1 | - | 5 | 1180 | |||
BC056221.1 | - | 3 | 767 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
