siRNA Details
Gene Symbol: Dnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173383.2 | NP_775559.2 | 3 | 318 | ||
GenBank mRNA | AK141343.1 | - | 3 | 303 | ||
AY321066.1 | - | 3 | 270 | |||
BC034897.1 | - | 2 | 653 | 3470697 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
