siRNA Details
Gene Symbol: Fstl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253719.1 | NP_001240648.1 | 9 | 1306 | ||
NM_178673.4 | NP_848788.2 | 9 | 1566 | |||
GenBank mRNA | AF374460.1 | - | 9 | 1540 | ||
AK033652.1 | - | 9 | 1557 | |||
AK046070.1 | - | 9 | 1307 | |||
AK047518.1 | - | 1 | 2189 | |||
AK081152.1 | - | 9 | 1557 | |||
AK138388.1 | - | 9 | 1858 | |||
BC075640.1 | - | 9 | 1529 | 30545529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
