siRNA Details
Gene Symbol: Fstl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253719.1 | NP_001240648.1 | 5, 6 | 773 | ||
NM_178673.4 | NP_848788.2 | 5, 6 | 1033 | |||
GenBank mRNA | AF374460.1 | - | 5, 6 | 1007 | ||
AK033652.1 | - | 5, 6 | 1024 | |||
AK046070.1 | - | 5, 6 | 774 | |||
AK081152.1 | - | 5, 6 | 1024 | |||
AK138388.1 | - | 5, 6 | 1325 | |||
AK140611.1 | - | 5, 6 | 1036 | |||
BC075640.1 | - | 5, 6 | 996 | 30545529 |

More Information
Other Species Targeted

Gene Ontology Categories:
