siRNA Details
Gene Symbol: Klhdc8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144810.5 | NP_659059.1 | 2 | 1085 | ||
GenBank mRNA | AK043039.1 | - | 2 | 423 | ||
AK044318.1 | - | 2 | 661 | |||
AK080356.1 | - | 2 | 1085 | |||
BC011135.1 | - | 2 | 650 | 4191503 | ||
BC023173.1 | - | 2 | 284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
