siRNA Details
Gene Symbol: Nudt18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153136.4 | NP_694776.2 | 3 | 992 | ||
GenBank mRNA | AK089446.1 | - | 2 | 780 | ||
AK137014.1 | - | 3 | 1206 | |||
AK145997.1 | - | 3 | 981 | |||
AK155086.1 | - | 3 | 993 | |||
AK157937.1 | - | 3 | 993 | |||
BC036718.1 | - | 3 | 1148 | 5322150 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
