siRNA Details
Gene Symbol: Plekhh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177606.4 | NP_808274.2 | 13, 14 | 2295 | ||
GenBank mRNA | AK029252.1 | - | 13, 14 | 2297 | ||
AK173337.1 | - | 10, 11 | 1950 | |||
BC125583.1 | - | 13, 14 | 2260 | 40130098 | ||
BC137804.1 | - | 13, 14 | 2257 | 8860823 |
