siRNA Details
Gene Symbol: Map9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081230.1 | NP_001074699.1 | 5 | 956 | ||
GenBank mRNA | AK030527.1 | - | 5 | 797 | ||
AK038737.1 | - | 5 | 901 | |||
AK135702.1 | - | 2 | 450 | |||
AK135823.1 | - | 5 | 860 | |||
AK138540.1 | - | 5 | 901 | |||
AK147287.1 | - | 5 | 899 | |||
AK147416.1 | - | 5 | 899 | |||
AK162555.1 | - | 5 | 876 | |||
BC061216.1 | - | 5 | 812 | |||
BC087935.1 | - | 5 | 853 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
