siRNA Details
Gene Symbol: Prepl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163622.1 | NP_001157094.1 | 9 | 1807 | ||
NM_001163623.1 | NP_001157095.1 | 8 | 1271 | |||
NM_001163624.1 | NP_001157096.1 | 8 | 1139 | |||
NM_145984.3 | NP_666096.3 | 9 | 1371 | |||
GenBank mRNA | AK028877.1 | - | 9 | 1765 | ||
AK050878.1 | - | 8 | 1133 | |||
AK083070.1 | - | 9 | 1312 | |||
AK129142.1 | - | 3 | 1225 | |||
BC004612.1 | - | 8 | 1225 | 3585402 |
