siRNA Details
Gene Symbol: Nutm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172521.1 | NP_766109.1 | 7 | 2969 | ||
GenBank mRNA | AK077049.1 | - | 7 | 2969 | ||
BC125332.1 | - | 7 | 2949 | 40129847 | ||
BC125334.1 | - | 7 | 2949 | 40129849 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
