siRNA Details
Gene Symbol: Sox30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173384.2 | NP_775560.1 | 2 | 1233 | ||
GenBank mRNA | AY005801.1 | - | 2 | 1233 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006246152.
Gene Ontology Categories:
