siRNA Details
Gene Symbol: A430105I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001982.2 | NP_001001982.2 | 9, 10 | 1072 | ||
GenBank mRNA | AK040533.1 | - | 8, 9 | 1113 | ||
AK137584.1 | - | 9, 10 | 1074 | |||
BC075684.1 | - | 8, 9 | 824 | 30611649 |
