siRNA Details
Gene Symbol: Nudt15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172527.2 | NP_766115.1 | 4 | 701 | ||
GenBank mRNA | AK032657.1 | - | 2 | 296 | ||
AK040749.1 | - | 4 | 704 | |||
AK043196.1 | - | 203 | ||||
AK168451.1 | - | 245 | ||||
BC060962.1 | - | 2 | 285 | 30302091 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
