siRNA Details
Gene Symbol: Gm4788 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029977.3 | NP_001025148.3 | 9, 10 | 1586 | ||
NM_001160303.1 | NP_001153775.1 | 9, 10 | 1583 | |||
NM_001160304.1 | NP_001153776.1 | 9, 10 | 1586 | |||
GenBank mRNA | AJ871401.1 | - | 9, 10 | 1515 | ||
AJ871402.1 | - | 9, 10 | 1515 | |||
BC141136.1 | - | 9, 10 | 1555 | 9055748 | ||
BC145444.1 | - | 9, 10 | 1555 | 9054002 |
