siRNA Details
Gene Symbol: PEX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000287.3 | NP_000278.3 | 12 | 2414 | ||
NM_001316313.1 | NP_001303242.1 | 12 | 2150 | |||
XM_011514661.1 | XP_011512963.1 | 11 | 2330 | |||
GenBank mRNA | AB051076.1 | - | 12 | 2321 | ||
AB051077.1 | - | 12 | 2057 | |||
AB208906.1 | - | 11 | 2510 | |||
AK314237.1 | - | 12 | 2350 | |||
BC033487.1 | - | 12 | 2329 | |||
BC048331.1 | - | 12 | 2337 | 5189685 | ||
D83703.1 | - | 12 | 2391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
