siRNA Details
Gene Symbol: Tmprss13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013373.2 | NP_001013391.2 | 11 | 1428 | ||
GenBank mRNA | BC010843.1 | - | 3 | 277 | ||
BC042878.1 | - | 10 | 1164 | |||
BC085323.1 | - | 11 | 1413 | 30734093 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
