siRNA Details
Gene Symbol: PEX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000287.3 | NP_000278.3 | 2 | 1096 | ||
NM_001316313.1 | NP_001303242.1 | 2 | 832 | |||
NR_133009.1 | 2 | 1096 | ||||
XM_011514661.1 | XP_011512963.1 | 2 | 1096 | |||
GenBank mRNA | AB051076.1 | - | 2 | 1003 | ||
AB051077.1 | - | 2 | 739 | |||
AB051078.1 | - | 2 | 1003 | |||
AB208906.1 | - | 2 | 1056 | |||
AK225704.1 | - | 2 | 1034 | |||
AK314237.1 | - | 2 | 1032 | |||
BC033487.1 | - | 2 | 1011 | |||
BC048331.1 | - | 2 | 1019 | 5189685 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
