siRNA Details
Gene Symbol: Gm136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033255.2 | NP_001028427.1 | 5 | 1118 | ||
GenBank mRNA | AK133458.1 | - | 5 | 1119 | ||
BC147519.1 | - | 5 | 1110 | 9056511 | ||
BC147522.1 | - | 5 | 1110 | 9056514 |
