siRNA Details
Gene Symbol: Sidt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289668.1 | NP_001276597.1 | 3 | 665 | ||
NM_172257.4 | NP_758461.1 | 3 | 665 | |||
GenBank mRNA | AK081177.1 | - | 3 | 651 | ||
AK152371.1 | - | 3 | 598 | |||
AK155875.1 | - | 3 | 569 | |||
AK196459.1 | - | 3 | 197 | |||
BC023957.1 | - | 3 | 669 | 5321190 | ||
BC051101.1 | - | 3 | 393 | 5052849 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
