siRNA Details
Gene Symbol: 4932411E22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172534.2 | NP_766122.1 | 3 | 430 | ||
GenBank mRNA | AK029964.1 | - | 3 | 431 | ||
BC140961.1 | - | 3 | 416 | 9055573 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
