siRNA Details
Gene Symbol: Arid5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172205.1 | NP_001165676.1 | 5 | 429 | ||
NM_001172206.1 | NP_001165677.1 | 5 | 426 | |||
NM_001290726.1 | NP_001277655.1 | 6 | 524 | |||
NM_001290727.1 | NP_001277656.1 | 4 | 305 | |||
NM_145996.4 | NP_666108.2 | 3 | 255 | |||
GenBank mRNA | AK029237.1 | - | 3 | 234 | ||
AK085015.1 | - | 3 | 235 | |||
AK149839.1 | - | 5 | 409 | |||
AK150170.1 | - | 6 | 504 | |||
AK155555.1 | - | 4 | 285 | |||
AK156376.1 | - | 5 | 409 | |||
AK156622.1 | - | 4 | 691 | |||
AK156934.1 | - | 2 | 1991 | |||
AK157607.1 | - | 5 | 409 | |||
BC027152.1 | - | 4 | 272 | 4935168 | ||
CX209955.1 | - | 4 | 288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
