siRNA Details
Gene Symbol: Lman2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013374.1 | NP_001013392.1 | 5 | 624 | ||
GenBank mRNA | AK041656.1 | - | 3 | 2469 | ||
AK142024.1 | - | 593 | ||||
AK158015.1 | - | 5 | 643 | |||
BC027226.1 | - | 4 | 542 | 2654285 | ||
BC046969.1 | - | 5 | 624 | 6313679 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
