siRNA Details
Gene Symbol: Rhot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145999.2 | NP_666111.1 | 17 | 1932 | ||
GenBank mRNA | AK142183.1 | - | 12 | 2038 | ||
AY311390.1 | - | 17 | 1932 | |||
BC023163.1 | - | 7 | 702 | |||
BC023927.1 | - | 3 | 985 | |||
BC029777.1 | - | 17 | 1565 | 4950061 | ||
BC034062.1 | - | 17 | 1587 | 4207654 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
