siRNA Details
Gene Symbol: Vezt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031848.1 | - | 9 | 1474 | ||
AK036017.1 | - | 9 | 1411 | |||
AK042540.1 | - | 9 | 1405 | |||
AY753561.1 | - | 9 | 1367 | |||
AY753562.1 | - | 9 | 1367 | |||
BC056428.1 | - | 9 | 1398 | 5716245 |

More Information
Other Species Targeted

