siRNA Details
Gene Symbol: Vgll2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300957.1 | NP_001287886.1 | 1, 2 | 311 | ||
NM_153786.2 | NP_722481.1 | 1, 2 | 311 | |||
GenBank mRNA | AF542181.1 | - | 1, 2 | 127 | ||
AJ578054.1 | - | 1, 2 | 129 | |||
AK048390.1 | - | 1, 2 | 283 | |||
AK052987.1 | - | 1, 2 | 286 | |||
BC133696.1 | - | 1, 2 | 127 | 40081288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
