siRNA Details
Gene Symbol: Trim50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178240.2 | NP_839971.1 | 2, 3 | 487 | ||
GenBank mRNA | AY081947.1 | - | 2, 3 | 487 | ||
BC130022.1 | - | 2, 3 | 490 | 40140742 |

More Information
Other Species Targeted

Gene Ontology Categories:
