siRNA Details
Gene Symbol: Rhbdd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146002.2 | NP_666114.1 | 3 | 707 | ||
GenBank mRNA | AK030434.1 | - | 4 | 763 | ||
AK032400.1 | - | 2 | 131 | |||
AK041845.1 | - | 3 | 653 | |||
AK043674.1 | - | 3 | 642 | |||
AK140006.1 | - | 3 | 745 | |||
AK141182.1 | - | 4 | 828 | |||
AK161545.1 | - | 1 | 2592 | |||
AK165926.1 | - | 3 | 671 | |||
BC018360.1 | - | 3 | 651 | 3980676 | ||
BC028957.1 | - | 3 | 629 | |||
BC053091.1 | - | 1 | 2273 | |||
BC098493.1 | - | 2 | 361 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
