siRNA Details
Gene Symbol: Il1f9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153511.3 | NP_705731.2 | 4 | 626 | ||
GenBank mRNA | AK081783.1 | - | 4 | 621 | ||
AK156687.1 | - | 623 | ||||
AY071843.1 | - | 3 | 352 | |||
BC116227.1 | - | 3 | 352 | 40091521 | ||
BC116228.1 | - | 3 | 352 | 40091522 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
