siRNA Details
Gene Symbol: Fcgbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122603.1 | NP_001116075.1 | 10 | 3560 | ||
GenBank mRNA | AJ011416.1 | - | 4 | 895 |
Gene Symbol: 9530053A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164655.1 | NP_001158127.1 | 10 | 3557 |

More Information
Other Species Targeted

Gene Ontology Categories:
