siRNA Details
Gene Symbol: Rap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024457.2 | NP_077777.1 | 6 | 560 | ||
GenBank mRNA | AK143910.1 | - | 6 | 543 | ||
AK153870.1 | - | 6 | 517 | |||
AK163242.1 | - | 6 | 624 | |||
AK167548.1 | - | 6 | 516 | |||
AK205627.1 | - | 2 | 89 | |||
BC033382.1 | - | 6 | 500 | |||
BC052480.1 | - | 6 | 560 | |||
BC094066.1 | - | 6 | 500 | |||
BC147390.1 | - | 6 | 432 | 9056464 | ||
BC147391.1 | - | 6 | 432 | 9056441 | ||
BC147700.1 | - | 5 | 418 | 9087459 | ||
BC147713.1 | - | 5 | 418 | 9087472 | ||
CK793274.1 | - | 1 | 77 |
