siRNA Details
Gene Symbol: Rap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024457.2 | NP_077777.1 | 3 | 277 | ||
GenBank mRNA | AK143910.1 | - | 3 | 260 | ||
AK153870.1 | - | 3 | 234 | |||
AK163242.1 | - | 3 | 341 | |||
AK167548.1 | - | 3 | 233 | |||
AK178034.1 | - | 3 | 150 | |||
AK196093.1 | - | 3 | 150 | |||
AK199293.1 | - | 3 | 151 | |||
AK218552.1 | - | 3 | 150 | |||
BC033382.1 | - | 3 | 217 | |||
BC052480.1 | - | 3 | 277 | |||
BC094066.1 | - | 3 | 217 | |||
BC147390.1 | - | 3 | 149 | 9056464 | ||
BC147391.1 | - | 3 | 149 | 9056441 | ||
BC147700.1 | - | 2 | 135 | 9087459 | ||
BC147713.1 | - | 2 | 135 | 9087472 | ||
BU848028.1 | - | 282 | ||||
M79314.1 | - | 1 | 67 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_134346, XM_008765377, U07795, BC081731, D88313, and others.
Gene Ontology Categories:
