siRNA Details
Gene Symbol: Prr14l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194340.2 | NP_919321.2 | 6 | 5225 | ||
GenBank mRNA | AY372183.1 | - | 1 | 446 | ||
BC094374.1 | - | 1 | 772 | 30531815 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
