siRNA Details
Gene Symbol: Rnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159624.1 | NP_001153096.1 | 9 | 1704 | ||
NM_145417.3 | NP_663392.2 | 10 | 1821 | |||
GenBank mRNA | AK031138.1 | - | 9 | 1644 | ||
AK145466.1 | - | 10 | 1825 | |||
AK159195.1 | - | 10 | 1764 | |||
AK159445.1 | - | 10 | 1753 | |||
AK159625.1 | - | 10 | 1764 | |||
BC010520.1 | - | 10 | 1782 | |||
BC019200.1 | - | 10 | 1792 | 5041005 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
