siRNA Details
Gene Symbol: Zbtb8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153541.3 | NP_705769.2 | 4 | 1267 | ||
GenBank mRNA | AK132152.1 | - | 4 | 1305 | ||
AK140914.1 | - | 4 | 1256 | |||
AK163930.1 | - | 4 | 1268 | |||
BC023839.1 | - | 4 | 1262 | 5344921 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
