siRNA Details
Gene Symbol: Arrdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162485.1 | NP_001155957.1 | 6 | 912 | ||
NM_178408.3 | NP_848495.2 | 6 | 909 | |||
GenBank mRNA | AK170018.1 | - | 6 | 786 | ||
BC004091.1 | - | 6 | 807 | |||
BC018501.1 | - | 6 | 791 | 3662534 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
