siRNA Details
Gene Symbol: Ccdc92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144819.2 | NP_659068.1 | 3 | 375 | ||
GenBank mRNA | AK028192.1 | - | 3 | 352 | ||
AK136203.1 | - | 3 | 541 | |||
AK165371.1 | - | 3 | 375 | |||
BC021492.1 | - | 3 | 319 | 5345674 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
