siRNA Details
Gene Symbol: BC021785 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001489.3 | NP_001001489.1 | 3 | 516 | ||
NM_001113385.1 | NP_001106856.1 | 3 | 516 | |||
NM_001113388.2 | NP_001106859.1 | 3 | 516 | |||
GenBank mRNA | AI527856.1 | - | 3 | 509 | ||
AK085266.1 | - | 3 | 517 | |||
AK165453.1 | - | 3 | 514 | |||
AK165493.1 | - | 3 | 513 | |||
BC021785.1 | - | 3 | 459 | |||
BC094014.1 | - | 2 | 243 | |||
BC147384.1 | - | 3 | 456 | |||
BC147385.1 | - | 3 | 456 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
